Learn More
Tocris Bioscience™ GW 501516
Potent and selective PPARδ agonist
Marca: Tocris Bioscience™ 5674/50
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
GW 501516 is a potent and selective PPARδ agonist (EC50 = 1.2 nM). Displays <1000-fold selectivity over other PPAR subtypes. Increases ABC A1 transporter expression and induces apolipoprotein A1-mediated cholesterol efflux in vitro. Also increase serum HDL cholesterol and lowers small, dense LDL levels in obesity in vivo models.Especificaciones
| 2-[2-Methyl-4-[[[4-methyl-2-[4-(trifluoromethyl)phenyl]-5-thiazolyl]methyl]thio]phenoxy]acetic acid | |
| 317318-70-0 | |
| 50 mg |