Learn More
GW 441756, MedChemExpress
Código de producto. 16433527
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
50 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16433527 50 mg 50 miligramos
16407508 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16433527 Proveedor Medchem Express N.º de proveedor HY18314
MedChemExpress GW 441756 is a potent and specific nerve growth factor (NGF) receptor tyrosine kinases A (TrkA) inhibitor (IC50=2 nM), which eliminates the BmK NSPK-induced neurite outgrowth.
GW 441756 is a potent and specific nerve growth factor (NGF) receptor tyrosine kinases A (TrkA) inhibitor (IC50=2 nM), which eliminates the BmK NSPK-induced neurite outgrowth.
Especificaciones
Nombre del producto químico o material GW 441756
CAS 504433-23-2
Color Yellow
Notas de grado de pureza Research
Fórmula molecular C17H13N3O
Cantidad 50 mg
Información de solubilidad DMSO : 17.5 mg/mL (63.57 mM; Need ultrasonic and warming)
SMILES O=C1/C(C2=NC=CC=C2N1)=C\C3=CN(C)C4=C3C=CC=C4
Peso molecular (g/mol) 275.3
Formula Weight (peso de la fórmula) 275.3
Porcentaje de pureza 97.14%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-Kinase/protease