missing translation for 'onlineSavingsMsg'
Learn More

Guggulsterone, MedChemExpress

Código de producto. 16480497
Encompass_Preferred
Change view
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
16480497 50 mg 50 miligramos
16474148 10 mg 10 miligramos
16456857 5 mg 5 miligramos
16435116 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 16480497 Proveedor Medchem Express N.º de proveedor HY107738

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

MedChemExpress Guggulsterone is a plant sterol derived from the gum resin of the tree Commiphora wightii.

Guggulsterone inhibits the growth of a wide variety of tumor cells and induces apoptosis through down regulation of antiapoptotic gene products (IAP1, xIAP, Bfl-1/A1, Bcl-2, cFLIP and survivin), modulation of cell cycle proteins (cyclin D1 and c-Myc), activation of caspases and JNK, inhibition of Akt. Guggulsterone, a farnesoid X receptor (FXR) antagonist, decreases CDCA-induced FXR activation with IC50s of 17 and 15 μM for Z- and E-Guggulsterone, respectively.

Specifications

Nombre del producto químico o material Guggulsterone
Forma física Solid
CAS 95975-55-6
Color Blanco
Cantidad 50 mg
Notas de grado de pureza Research
Fórmula molecular C21H28O2
Para utilizar con (aplicación) Cancer-Kinase/protease
Sinónimo Z/E-Guggulsterone
Información de solubilidad DMSO : 20.83 mg/mL (66.67 mM; Need ultrasonic)
SMILES C[C@@]1(/C2=C/C)[C@](CC2=O)([H])[C@@]3([H])[C@]([C@@]4(C(CC3)=CC(CC4)=O)C)([H])CC1
Peso molecular (g/mol) 312.45
Formula Weight (peso de la fórmula) 312.45
Porcentaje de pureza 99.5%
Grado Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.