Learn More
GSK963, MedChemExpress
Código de producto. 16493778
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
Tamaño de la unidad:
10 miligramos
25 miligramos
5 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16493778 10 mg 10 miligramos
16436507 5 mg 5 miligramos
16412856 25 mg 25 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16493778 Proveedor Medchem Express N.º de proveedor HY103028A
MedChemExpress GSK963 is a chiral, highly potent and selective inhibitor of RIP1 kinase, with an IC50 of 29 nM. GSK963 is a selective and potent inhibitor of necroptosis in murine and human cells in vitro.
GSK963 is a chiral, highly potent and selective inhibitor of RIP1 kinase, with an IC50 of 29 nM. GSK963 is a selective and potent inhibitor of necroptosis in murine and human cells in vitro.
Especificaciones
Nombre del producto químico o material GSK963
Forma física Solid
CAS 2049868-46-2
Color Amarillo claro
Cantidad 10 mg
Notas de grado de pureza Research
Fórmula molecular C14H18N2O
Información de solubilidad DMSO : 25 mg/mL (108.55 mM; Need ultrasonic)
SMILES CC(C)(C)C(N1N=CC[C@H]1C2=CC=CC=C2)=O
Peso molecular (g/mol) 230.31
Formula Weight (peso de la fórmula) 230.31
Porcentaje de pureza 99.15%