Learn More
GSK805, MedChemExpress
Código de producto. 16475338
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16475338 10 mg 10 miligramos
16448057 5 mg 5 miligramos
16441537 50 mg 50 miligramos
16435976 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16475338 Proveedor Medchem Express N.º de proveedor HY12776
MedChemExpress GSK805 is an orally active and CNS penetrant RORγt inhibitor. GSK805 inhibits RORγ and Th17 cells differentiation with pIC50 values of 8.4 and >8.2. GSK805 inhibits the function of Th17 cells. GSK805 can be used for the research of immunity.
GSK805 is an orally active and CNS penetrant RORγt inhibitor. GSK805 inhibits RORγ and Th17 cells differentiation with pIC50 values of 8.4 and >8.2. GSK805 inhibits the function of Th17 cells. GSK805 can be used for the research of immunity.
Especificaciones
Nombre del producto químico o material GSK805
CAS 1426802-50-7
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C23H18Cl2F3NO4S
Cantidad 10 mg
Información de solubilidad DMSO : ≥ 100 mg/mL (187.84 mM)
SMILES O=C(NC1=CC(Cl)=C(C2=CC=CC=C2OC(F)(F)F)C(Cl)=C1)CC3=CC=C(S(=O)(CC)=O)C=C3
Peso molecular (g/mol) 532.36
Formula Weight (peso de la fórmula) 532.36
Porcentaje de pureza 98.26%
Grado Research
Forma física Solid
Para utilizar con (aplicación) COVID-19-immunoregulation