Learn More
GSK2850163, MedChemExpress
Código de producto. 16424126
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
Tamaño de la unidad:
10 miligramos
25 miligramos
5 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16424126 25 mg 25 miligramos
16412668 5 mg 5 miligramos
16400469 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16424126 Proveedor Medchem Express N.º de proveedor HYU00459
MedChemExpress GSK2850163 is a novel inhibitor of inositol-requiring enzyme-1 alpha (IRE1α) which can inhibit IRE1α kinase activity and RNase activity with IC50s of 20 and 200 nM, respectively.
GSK2850163 is a novel inhibitor of inositol-requiring enzyme-1 alpha (IRE1α) which can inhibit IRE1α kinase activity and RNase activity with IC50s of 20 and 200 nM, respectively.
Especificaciones
Nombre del producto químico o material GSK2850163
CAS 2121989-91-9
Color Light Yellow
Notas de grado de pureza Research
Fórmula molecular C24H29Cl2N3O
Cantidad 25 mg
Información de solubilidad DMSO : 67.5 mg/mL (151.21 mM; Need ultrasonic) ∣Ethanol : 50 mg/mL (112.00 mM; Need ultrasonic)
SMILES O=C(N(CCC1)C[C@]21CCN(CC3=CC=C(Cl)C(Cl)=C3)C2)NCC4=CC=C(C)C=C4
Peso molecular (g/mol) 446.41
Formula Weight (peso de la fórmula) 446.41
Porcentaje de pureza 98.0%
Grado Research
Forma física Oil
Para utilizar con (aplicación) Cancer-Kinase/protease