Learn More
GSK163090, MedChemExpress
Código de producto. 16492177
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16492177 50 mg 50 miligramos
16486446 100 mg 100 miligramos
16486038 10 mg 10 miligramos
16448647 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16492177 Proveedor Medchem Express N.º de proveedor HY14348
MedChemExpress GSK163090 is a potent, selective and orally active 5-HT1A/1B/1D receptor antagonist with pKi values of 9.4/8.5/9.7, respectively. GSK163090 inhibits the functional activity of serotonin reuptake transporter (SerT) with a pKi value of 6.1. GSK163090 has antidepressant and anxiolytic activities.
GSK163090 is a potent, selective and orally active 5-HT1A/1B/1D receptor antagonist with pKi values of 9.4/8.5/9.7, respectively. GSK163090 inhibits the functional activity of serotonin reuptake transporter (SerT) with a pKi value of 6.1. GSK163090 has antidepressant and anxiolytic activities.
Especificaciones
Nombre del producto químico o material GSK163090
CAS 844903-58-8
Color Off-White
Notas de grado de pureza Research
Fórmula molecular C25H29N5O
Cantidad 50 mg
Información de solubilidad DMSO : 14.29 mg/mL (34.39 mM; Need ultrasonic)
SMILES O=C1NCCN1C2=CC=CC(CCN3CCN(C4=C5C=CC(C)=NC5=CC=C4)CC3)=C2
Peso molecular (g/mol) 415.53
Formula Weight (peso de la fórmula) 415.53
Porcentaje de pureza 98.96%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Neuroscience-Neuromodulation