Learn More
Tocris Bioscience™ GSK 2833503A
Descripción
GSK 2833503A is a potent and selective TRPC6 and TRPC3 antagonist (IC50 = 3-16 nM and 21-100 nM, respectively). GSK 2833503A exhibits >63-fold selectivity over other ion channels, including other TRP channels, CaV1.2, hERG and NaV1.5. GSK 2833503A suppresses angiotensin II or endothelin-1-induced cardiac hypertrophy signaling in HEK293 cells in vitro and in cardiomyocytes.
Especificaciones
Especificaciones
| Nombre del producto químico o material | [2-[(4-Chloro-2-fluorophenyl)amino]-5-methyl-4-thiazolyl][(2 S,3 S)-2,3-dimethyl-1-piperidinyl]methanone |
| CAS | 1366234-01-6 |
| Cantidad | 10 mg |
| Objetivo | TRPC Channel Blockers |
| Fórmula molecular | C18H21ClFN3OS |
| Pureza | 0.98 |