Learn More
Tocris Bioscience™ GSK 1702934A
Descripción
GSK 1702934A is a potent and selective TRPC3/6 activator (EC50 = 80 and 440 nM for TRPC3 and 6, respectively). Exhibits no activity at TRPV4, TRPA1, M1, M4, CaV1.2, hERG, NaV1.5, or CXCR5 receptors at concentrations <10 μmol/L. Induces TRPC3/6-currents in HEK293 cells transduced with recombinant human TRPC3/6. Directly activates TRPC channels through by-passing phospholipase-C signaling. Also induces calcium signals in HEK-TRPC6 cells (pEC50 = 6.6).
Especificaciones
Especificaciones
| Nombre del producto químico o material | 1,3-Dihydro-1-[1-[(5,6,7,8-tetrahydro-4H-cyclohepta[b]thien-2-yl)carbonyl]-4-piperidinyl]-2H-benzimidazol-2-one |
| CAS | 924377-85-5 |
| Cantidad | 10 mg |
| Fórmula molecular | C22H25N3O2S |
| Formula Weight (peso de la fórmula) | 395.52 |
| Pureza | 98% |