Learn More
GR 203040, Tocris Bioscience™
GREENER_CHOICE
Potent, selective NK1 receptor antagonist
Marca:  Tocris Bioscience 4647/5
Encompass
Código de producto. 15917900
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos »
Descripción
Descripción
GR 203040 is a potent and selective NK1 receptor antagonist. Potently inhibits substance P binding to NK1 receptors (pKi = 10.1 to 10.5). Displays antiemetic activity in vivo.
Especificaciones
Especificaciones
168398-02-5
(2S,3S)-N-[[2-Methoxy-5-(1H-tetrazol-1-yl)phenyl]methyl]-2-phenyl-3-piperidinamine dihydrochloride