Learn More
Tocris Bioscience™ GR 203040
Descripción
GR 203040 is a potent and selective NK1 receptor antagonist. Potently inhibits substance P binding to NK1 receptors (pKi = 10.1 to 10.5). Displays antiemetic activity in vivo.
Especificaciones
Especificaciones
| CAS | 168398-02-5 |
| Sinónimo | (2S,3S)-N-[[2-Methoxy-5-(1H-tetrazol-1-yl)phenyl]methyl]-2-phenyl-3-piperidinamine dihydrochloride |
| Cantidad | 5 mg |
| Fórmula molecular | C20H24N6O.2HCl |
| Formula Weight (peso de la fórmula) | 437.37 |
| Pureza | 98% |