missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gr-1 (Ly-6G/Ly-6C) Rat, anti-Mouse, PE, Clone: RB6-8C5, STEMCELL Technologies
Descripción
- The RB6-8C5 antibody reacts with the structurally related murine Ly-6G (21 - 25 kDa) and Ly-6C (14 - 16 kDa) GPI-anchored proteins, which together comprise the granulocyte receptor-1 antigen (Gr-1)
- Gr-1 is expressed on monocytes, neutrophils, subsets of macrophages, plasmacytoid dendritic cells, and T cells
- Monocytes in the bone marrow transiently express Gr-1 during development and the expression level is strongly correlated with granulocyte differentiation and maturation
- In the periphery Gr-1 is found predominantly on neutrophils and is a useful marker for these cells
- Whereas the RB6-8C5 binds to both Ly-6G and Ly-6C, another commonly used anti-Gr-1 antibody, clone 1A8, binds specifically to Ly-6G
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse Neutrophil Enrichment Kit (Catalog #19762)
Especificaciones
Especificaciones
| Antígeno | Gr-1 (Ly-6G/Ly-6C) |
| Aplicaciones | Immunoprecipitation, Western Blot, Flow Cytometry, Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | RB6-8C5 |
| Conjugado | PE |
| Especie del huésped | Rat |
| Cantidad | 50 μg |
| Primario o secundario | Primary |
| Especies diana | Mouse |
| Isotype | IgG2b κ |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?