missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPR176 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15712546
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15712546 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15712546 Proveedor Invitrogen™ N.º de proveedor PA3046

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

GPR176 is in the G protein-coupled receptor family and are cell surface receptors involved in responses to hormones, growth factors, and neurotransmitters. Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Hamster, Elephant, Bovine, Dog, Bat, Horse, Stickleback, Zebrafish (100%). GPR176 exists in different glycosylated forms.

G protein-coupled receptors (GPRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, comprise a superfamily of proteins that play a role in many different stimulus-response pathways. G protein coupled receptors translate extracellular signals into intracellular signals (G protein activation) and they respond to a variety of signaling molecules, such as hormones and neurotransmitters. GPR176 (G protein-coupled receptor 176), also known as HB-954, GPR or Gm1012, is a 515 amino acid multi-pass membrane protein belonging to the G-protein coupled receptor 1 family. Expressed in brain and spleen, with trace expression in kidney, GPR176 functions as an orphan receptor that is thought to play a role in signaling events throughout the cell. Containing four N-glycosylation sites, seven transmembrane domains and a large C-terminal cytosolic domain, GPR176 is encoded by a gene mapping to human chromosome 15q14.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno GPR176
Aplicaciones Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración Conc. Not Determined
Conjugado Unconjugated
Formulación Whole Serum with 0.05% sodium azide
génica GPR176
N.º de referencia del gen Q14439, Q80WT4
Alias de gen Agr9; G protein-coupled receptor 176; Gm1012; Gpr; GPR176; G-protein coupled receptor 176; G-protein coupled receptor AGR9; HB954; HB-954; probable G-protein coupled receptor 176; putative G protein coupled receptor; RBU-15
Símbolos de los genes GPR176
Especie del huésped Rabbit
Inmunógeno KLH conjugated COOH-terminal amino acids of 496-515 (RVERKMSRNNKVSIFPKVDS).
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 11245, 381413
Especies diana Human, Mouse
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.