missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPR158 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15720981 Tienda Thermo Scientific Productos
Change view
Click to view available options
Cantidad:
100 μL
Unit Size:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
15720981 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15720981 Supplier Invitrogen™ Supplier No. PA553030

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LGLAGKTQTA GVEERTKSQK PLPKDKETNR NHSNSDNTET KDPAPQNSNP AEEPRKPQKS GIMKQQRVNP TTANSDLNPG TTQMKDNFDI GEVCPWEVYD LTPGPVPSES KVQKHVSIVA SEMEKNPTFS LKEKSHHKP Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 76%.

Orphan receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno GPR158
Aplicaciones Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 0.1 mg/mL
Conjugado Unconjugated
Formulación PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
génica GPR158
N.º de referencia del gen Q5T848
Alias de gen 5330427M13Rik; G protein-coupled receptor 158; Gpr158; KIAA1136; mKIAA1136; Probable G-protein coupled receptor 158
Símbolos de los genes GPR158
Especie del huésped Rabbit
Inmunógeno Recombinant protein corresponding to Human GPR158. Recombinant protein control fragment (Product #RP-90844).
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 57512
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.