Learn More
GPR116 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16852514 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
20 μL
150 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16852514 20 μL 20 microlitros
16842514 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16852514 Proveedor Proteintech N.º de proveedor 140471AP20UL
Rabbit Polyclonal Antibody
G protein-coupled receptors (GPRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, comprise a superfamily of proteins that play a role in many different stimulus-response pathways. G protein coupled receptors translate extracellular signals into intracellular signals (G protein activation) and they respond to a variety of signaling molecules, such as hormones and neurotransmitters. GPR116 (G protein-coupled receptor 116) is a 1,346 amino acid multi-pass membrane protein that contains one SEA domain, one GPS domain and three Ig-like domains and belongs to the GPR family. Existing as a dilsulfide-liked homodimer at the cell surface, GPR116 exists as multiple alternatively spliced isoforms and is thought to play a role in regulating and maintaining proper acid-base balance throughout the cell.
Especificaciones
Antígeno GPR116
Aplicaciones Western Blot, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 0.25 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica ADGRF5
N.º de referencia del gen G5E8Q8, Q8IZF2, Q9WVT0
Alias de gen G protein coupled receptor 116, GPR116, KIAA0758, KPG_001
Símbolos de los genes ADGRF5
Especie del huésped Rabbit
Inmunógeno GPR116 Fusion Protein Ag5214
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 221395, 224792, 245977
Especies diana Human, Rat, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG