Learn More
Tocris Bioscience™ GP 1a
Descripción
GP 1a is a CB2 receptor inverse agonist (pEC50 = 7.1), shown in cAMP, GTPγS and β-arrestin recruitment assays. Demonstrates approximately 30-fold selectivity for CB2 receptors over CB1 receptors. Increases P-ERK1/2 expression in HL-60 cells in vitro.
Especificaciones
Especificaciones
| Nombre del producto químico o material | N-(Piperidin-1-yl)-1-(2,4-dichlorophenyl)-1,4-dihydro-6-methylindeno[1,2-c]pyrazole-3-carboxamide |
| CAS | 511532-96-0 |
| Cantidad | 10 mg |
| Objetivo | Cannabinoid R2 / CB2 Inverse Agonists |
| Fórmula molecular | C23H22Cl2N4O |
| Pureza | 0.99 |