Learn More
Invitrogen™ Goat anti-Human IgG (Kappa light chain) Recombinant Secondary Antibody, HRP
Código de producto. 17876082
Change view
Click to view available options
Cantidad:
50 μL
250 μL
Tamaño de la unidad:
250 microlitros
50 microlitros
17876082 250 μL 250 microlitros
17857482 50 μL 50 microlitros
2 options
Código de producto. 17876082
Marca: Invitrogen™ A56864
Goat Recombinant Monoclonal Secondary Antibody
This Goat-anti-Human IgG (Kappa Light Chain), Recombinant Secondary Antibody specifically detects human IgG Kappa Light chain with no cross-reactivity to Lambda light chain, heavy chain, and other species immunoglobulins. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include better specificity, animal origin-free formulation, and lot-to-lot consistency. The sensitivity and specificity of each lot is confirmed using ELISA. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Human IgG (Kappa light chain)
Aplicaciones ELISA, Western Blot
Clasificación Recombinant Monoclonal
Clon 81H59L2
Concentración 1.0 mg/mL
Conjugado HRP
Formulación PBS with proprietary stabilizer and proprietary preservative; pH 7.4
Especie del huésped Goat
Inmunógeno Purified Human IgG.
Método de purificación Protein G
Cantidad 250 μL
Estado normativo RUO
Primario o secundario Secondary
Especies diana Human
Contenido y almacenamiento 4°C, do not freeze
Tipo de producto Secondary Antibody
Formulario Liquid
Isotype IgG