missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: MEEWDVPQMK KEVESLKYQL AFQREMASKT IPELLKWIED GIPKDPFLNP DLMKNNPWVE KGKCTIL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
In mammals, G protein α, β and γ polypeptides are encoded by at least 16, 4 and 7 genes, respectively. Most interest in G proteins has been focused on their α subunits, since these proteins bind and hydrolyze GTP and most obviously regulate the activity of the best studied effectors. Evidence, however, has established an important regulatory role for the βγ subunits. It is becoming increasingly clear that different G protein complexes expressed in different tissues carry structurally distinct members of the γ as well as the α and β subunits, and that preferential associations between members of subunit families increase G protein functional diversity.
Spécification
Spécification
| Antígeno | GNG13 |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Concentración | 0.20 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | GNG13 |
| N.º de referencia del gen | Q9P2W3 |
| Alias de gen | 1500031D04Rik; AB030194; G gamma subunit, clone 2-35; G gamma subunit, clone:h2-35; G protein subunit gamma 13; G(gamma)13; Ggamma13; Gng13; guanine nucleotide binding protein (G protein), gamma 13; guanine nucleotide binding protein 13, gamma; guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-13; h2-35 |
| Símbolos de los genes | GNG13 |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?
