Learn More
GNE-3511, MedChemExpress
Código de producto. p-200098117
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16495438 10 mg 10 miligramos
16491617 50 mg 50 miligramos
16456036 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16495438 Proveedor Medchem Express N.º de proveedor HY12947
MedChemExpress GNE-3511 is an orally active bioavailable and brain-penetrant dual leucine zipper kinase (DLK) inhibitor with a Ki of 0.5 nM. GNE-3511 can cross the blood-brain-barrier and can be used for the research of neurodegenerative diseases.
GNE-3511 is an orally active bioavailable and brain-penetrant dual leucine zipper kinase (DLK) inhibitor with a Ki of 0.5 nM. GNE-3511 can cross the blood-brain-barrier and can be used for the research of neurodegenerative diseases.
Especificaciones
Nombre del producto químico o material GNE-3511
CAS 1496581-76-0
Color Light Yellow
Notas de grado de pureza Research
Fórmula molecular C23H26F2N6O
Cantidad 10 mg
Información de solubilidad DMSO : 31.25 mg/mL (70.94 mM; Need ultrasonic)
SMILES N#CC1=CC=NC(NC2=NC(N3CC(F)(F)CC3)=CC(C4CCN(C5COC5)CC4)=C2)=C1
Peso molecular (g/mol) 440.49
Formula Weight (peso de la fórmula) 440.49
Porcentaje de pureza 98.87%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-Kinase/protease