missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GNAS (R201C Mutant) Polyclonal Antibody
GREENER_CHOICE

Código de producto. 17790954
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17790954 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17790954 Proveedor Invitrogen™ N.º de proveedor PA5119556

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

This locus has a highly complex imprinted expression pattern. It gives rise to maternally, paternally, and biallelically expressed transcripts that are derived from four alternative promoters and 5' exons. Some transcripts contain a differentially methylated region (DMR) at their 5' exons, which is commonly found in imprinted genes and correlates with transcript expression. This gene has an antisense transcript. One of the transcripts produced from this locus, and the antisense transcript, are both paternally expressed noncoding RNAs, and may regulate imprinting in this region. In addition, one of the transcripts contains a second overlapping ORF, which encodes a structurally unrelated protein - Alex. Alternative splicing of downstream exons is also observed, which results in different forms of the stimulatory G-protein alpha subunit, a key element of the classical signal transduction pathway linking receptor-ligand interactions with the activation of adenylyl cyclase and a variety of cellular reponses. Additional transcript variants have been found for this gene, but the full-length nature and/or biological validity of some variants have not been determined. [provided by RefSeq, Jun 2015].
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno GNAS (R201C Mutant)
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.96 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
génica GNAS
N.º de referencia del gen O95467, P63092, P63094, P84996, Q5JWF2, Q6R0H6, Q6R0H7, Q9Z0F1
Alias de gen 5530400H20Rik; A930027G11Rik; alpha stimulating) complex locus; C130027O20Rik; Galphas; GNAS (guanine nucleotide binding protein; Gnas1; Gnasxl; GPSA; Gsa; Gs-alpha; GSP; Nesp; Nesp55; Nespl; Oedsml; Oed-Sml; P1; P2; P3; PHP1A; PHP1B; POH; SCG6
Símbolos de los genes GNAS
Especie del huésped Rabbit
Inmunógeno Carrier-protein conjugated synthetic peptide surrounding mutant R201C of mouse GNAS. The exact sequence is proprietary.
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 14683, 2778
Especies diana Human, Mouse
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.