Learn More
Glycochenodeoxycholic acid, MedChemExpress
Código de producto. 16439288
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
10 mg
Tamaño de la unidad:
10 miligramos
16439288 10 mg 10 miligramos
1 options
Código de producto. 16439288 Proveedor Medchem Express N.º de proveedor HYN233410mg
MedChemExpress Glycochenodeoxycholic acid (Chenodeoxycholylglycine) is a bile acid formed in the liver from chenodeoxycholate and glycine. It acts as a detergent to solubilize fats for absorption and is itself absorbed. Glycochenodeoxycholic acid (Chenodeoxycholylglycine) induces hepatocyte apoptosis.
Glycochenodeoxycholic acid (Chenodeoxycholylglycine) is a bile acid formed in the liver from chenodeoxycholate and glycine. It acts as a detergent to solubilize fats for absorption and is itself absorbed. Glycochenodeoxycholic acid (Chenodeoxycholylglycine) induces hepatocyte apoptosis.
Especificaciones
Nombre del producto químico o material Glycochenodeoxycholic acid
CAS 640-79-9
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C26H43NO5
Cantidad 10 mg
Sinónimo Chenodeoxycholylglycine
Información de solubilidad DMSO : 100 mg/mL (222.41 mM; Need ultrasonic)
SMILES C[C@@]12[C@](CC[C@]2([H])[C@H](C)CCC(NCC(O)=O)=O)([H])[C@@]3([H])[C@]([C@@]4([C@](C[C@H](O)CC4)([H])C[C@H]3O)C)([H])CC1
Peso molecular (g/mol) 449.62
Formula Weight (peso de la fórmula) 449.62
Porcentaje de pureza 98.0%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-programmed cell death