Learn More
Tocris Bioscience™ GLP-2 (3-33)
Descripción
GLP-2(3-33) is a peptide antagonist of glucagon-like peptide-2 (GLP-2) receptor. In cells, GLP-2(3-33) is generated by cleaving off the two N-terminal amino acids of GLP-2 with dipeptidylpeptidase IV (DPPIV). In high-fat diet (HFD) fed mice, GLP-2(3-33) treatment results in increased dyslipidemia and hepatic lipid accumulation. Chronic treatment of GLP-2(3-33) in HFD fed mice can cause hyperglycemia, glucose intolerance, high plasma insulin level after glucose load, increased pancreas weight and β-cell expansion.
Especificaciones
Especificaciones
| Gene ID (Entrez) | 9340 |
| Pureza | 0.95 |
| CAS | 275801-62-2 |
| Contenido y almacenamiento | Store at -20°C |
| Alias de gen | GLP-2 receptor, GLP-2 R, GLP-2-R, glucagon-like peptide 2 receptor |
| Peso molecular | M.W.-observed: 3557.93 |
| Cantidad | 1 mg |