Learn More
Kova Plastics by Alltrista Glasstic™ Slide 10 with Grids
Código de producto. 15292826
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
100
Tamaño de la unidad:
paquete de 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad Tamaño de la unidad
15292826 100 paquete de 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Código de producto. 15292826 Proveedor Kova Plastics by Alltrista N.º de proveedor 87144E
 Artículo restringido
Este artículo requiere una actualización de precios antes de ser solicitado.
Solicite el precio de este artículo aquí.
missing translation for 'validMainframeMsg'
A precision manufactured, 10-chambered disposable microscope slide made of optically clear plastic with a Neubauer counting grid.
  • The slide grid is in the focal plane of the specimen, allowing for use over a range of magnifications
  • Reduces pipetting errors and eliminates variables that can interfere with microscopic urinalysis
  • Superior optical plastic with a refractive index that is similar to glass
  • Manufactured to the specifications of high quality optical lenses, smooth with a tolerance below 50
  • Perimeter contained wells for precise volume control and protection against cross-contamination and leakage
  • Easier and faster counting with Neubauer-style ruling pattern
  • Capillary action provides reproducible precision
  • Homogeneous suspension of sample sediments
Especificaciones
Frosted Not Frosted
Material Optical Plastic
Tipo Slide with Counting Grid
Número de pocillos 10
Color Transparent
Descripción Neubauer style ruling pattern
Para utilizar con (aplicación) Microscopy applications
Cantidad 100