Learn More
Ginsenoside Rk3, MedChemExpress
Código de producto. 16409188
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
Tamaño de la unidad:
10 miligramos
5 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16409188 10 mg 10 miligramos
16401598 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16409188 Proveedor Medchem Express N.º de proveedor HYN0906
MedChemExpress Ginsenoside Rk3 is present in the roots Panax notoginseng herbs. Ginsenoside Rk3 significantly inhibits TNF-α-induced NF-κB transcriptional activity, with an IC50 of 14.24±1.30 μM in HepG2 cells.
Ginsenoside Rk3 is present in the roots Panax notoginseng herbs. Ginsenoside Rk3 significantly inhibits TNF-α-induced NF-κB transcriptional activity, with an IC50 of 14.24±1.30 μM in HepG2 cells.
Especificaciones
Nombre del producto químico o material Ginsenoside Rk3
CAS 364779-15-7
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C36H60O8
Cantidad 10 mg
Información de solubilidad DMSO : 100 mg/mL (161.07 mM; Need ultrasonic)
SMILES C[C@@]([C@@]12C)(C[C@H](O[C@]([C@@H]([C@@H](O)[C@@H]3O)O)([H])O[C@@H]3CO)[C@@]4([H])C5(C)C)[C@@](C[C@@H](O)[C@]1([H])[C@@H](C(CC/C=C(C)/C)=C)CC2)([H])[C@]4(CC[C@@H]5O)C
Peso molecular (g/mol) 620.86
Formula Weight (peso de la fórmula) 620.86
Porcentaje de pureza 98.85%
Grado Research
Forma física Solid
Para utilizar con (aplicación) COVID-19-immunoregulation