Learn More
Ginsenoside Rg2, MedChemExpress
Código de producto. 16419038
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
Tamaño de la unidad:
10 miligramos
5 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16419038 10 mg 10 miligramos
16401458 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16419038 Proveedor Medchem Express N.º de proveedor HYN0602
MedChemExpress Ginsenoside Rg2 is one of the major active components of ginseng. Ginsenoside Rg2 inhibits VCAM-1 and ICAM-1 expressions stimulated with lipopolysaccharide (LPS). Ginsenoside Rg2 also reduces Aβ1-42 accumulation.
Ginsenoside Rg2 is one of the major active components of ginseng. Ginsenoside Rg2 inhibits VCAM-1 and ICAM-1 expressions stimulated with lipopolysaccharide (LPS). Ginsenoside Rg2 also reduces Aβ1-42 accumulation.
Especificaciones
Nombre del producto químico o material Ginsenoside Rg2
CAS 52286-74-5
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C42H72O13
Cantidad 10 mg
Sinónimo Chikusetsusaponin I Panaxoside Rg2 Prosapogenin C2
Información de solubilidad DMSO : 100 mg/mL (127.39 mM; ultrasonic and warming and heat to 60°C)
SMILES C[C@]([C@@](C[C@H]1O)([H])[C@]2(CC[C@@H]3O)C)(C[C@H](O[C@@](O[C@H](CO)[C@@H](O)[C@@H]4O)([H])[C@@H]4O[C@@](O[C@@H](C)[C@H](O)[C@H]5O)([H])[C@@H]5O)[C@@]2([H])C3(C)C)[C@]6([C@@]1([H])[C@]([C@@](C)(O)CC/C=C(C)/C)([H])CC6)C
Peso molecular (g/mol) 785.01
Formula Weight (peso de la fórmula) 785.01
Porcentaje de pureza 98.0%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-programmed cell death