Learn More
Ginsenoside Rb1, MedChemExpress
Descripción
Ginsenoside Rb1, a main constituent of the root of Panax ginseng, inhibits Na+, K+-ATPase activity with an IC50 of 6.3±1.0 μM. Ginsenoside also inhibits IRAK-1 activation and phosphorylation of NF-κB p65 .
Specifications
Specifications
| Nombre del producto químico o material | Ginsenoside Rb1 |
| CAS | 41753-43-9 |
| Color | Blanco |
| Notas de grado de pureza | Research |
| Fórmula molecular | C54H92O23 |
| Cantidad | 5 mg |
| Sinónimo | Gypenoside III |
| Información de solubilidad | DMSO : 100 mg/mL (90.15 mM; Need ultrasonic) |
| SMILES | C[C@@]([C@@]12C)(CC[C@@]3([H])C4(C)C)[C@@](C[C@@H](O)[C@]1([H])[C@]([C@@](CC/C=C(C)/C)(C)O[C@@H]([C@@H]([C@@H](O)[C@@H]5O)O)O[C@@H]5CO[C@@H]([C@@H]([C@@H](O)[C@@H]6O)O)O[C@@H]6CO)([H])CC2)([H])[C@]3(CC[C@@H]4O[C@@](O[C@H](CO)[C@@H](O)[C@@H]7O)([H])[C@@H]7O[C@]([C@@H]([C@@H](O)[C@@H]8O)O)([H])O[C@@H]8CO)C |
| Peso molecular (g/mol) | 1109.29 |
| Show More |