Learn More
MedChemExpress Gigantol
Código de producto. 16444947
Encompass_Preferred
Cambiar vista
Click to view available options
50 mg
Código de producto.
16444947 50 mg
1 options
Código de producto. 16444947 Proveedor MedChemExpress N.º de proveedor HYN2523
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos
MedChemExpress Gigantol is a bibenzyl compound derived from several medicinal orchids. Giganto shows promising therapeutic potential against cancer cells. Gigantol is a novel inhibitor of the Wnt/β-catenin pathway.
Especificaciones
Contenido y almacenamiento Pure form -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Peso molecular 274.31 g/mol
Condiciones de envío Room temperature
Para utilizar con (aplicación) Cancer-programmed cell death
CAS 67884-30-4
Tipo de producto Natural Products
Pureza 99.79%
Solubilidad DMSO : ≥ 100 mg/mL (364.55 mM) ∣Ethanol : 50 mg/mL (182.28 mM∣Need ultrasonic)
Objetivo Wnt