Learn More
Gemcabene calcium, MedChemExpress
Código de producto. 16444308
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16444308 10 mg 10 miligramos
16437027 5 mg 5 miligramos
16435226 100 mg 100 miligramos
16420637 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16444308 Proveedor Medchem Express N.º de proveedor HY109567A
MedChemExpress Gemcabene calcium (PD-72953 calcium), a first-in-class lipid-lowering agent, lowers low-density lipoprotein cholesterol (LDL-C), decreases triglycerides, and raises high-density lipoprotein cholesterol (HDL-C) and lowers pro-inflammatory acute-phase protein, C-reactive protein (CRP), exerting anti-inflammatory activity.
Gemcabene calcium (PD-72953 calcium), a first-in-class lipid-lowering agent, lowers low-density lipoprotein cholesterol (LDL-C), decreases triglycerides, and raises high-density lipoprotein cholesterol (HDL-C) and lowers pro-inflammatory acute-phase protein, C-reactive protein (CRP), exerting anti-inflammatory activity.
Especificaciones
Nombre del producto químico o material Gemcabene calcium
Forma física Solid
CAS 209789-08-2
Color Blanco
Cantidad 10 mg
Notas de grado de pureza Research
Fórmula molecular C16H28CaO5
Para utilizar con (aplicación) COVID-19-immunoregulation
Sinónimo PD-72953 calcium
Información de solubilidad H2O : 10 mg/mL (29.37 mM; Need ultrasonic)
SMILES O=C([O-])C(C)(C)CCCCOCCCCC(C)(C([O-])=O)C.[Ca+2]
Peso molecular (g/mol) 340.47
Formula Weight (peso de la fórmula) 340.47
Porcentaje de pureza 98.0%