Learn More
Gefarnate, MedChemExpress
Código de producto. 16424657
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
50 mg
100 mg
Tamaño de la unidad:
100 miligramos
50 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16424657 50 mg 50 miligramos
16408876 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16424657 Proveedor Medchem Express N.º de proveedor HYB2206
MedChemExpress Gefarnate is a drug used for the treatment of gastritis and gastric ulcer, and has been proposed for use in the treatment of dry eye syndrome.
Gefarnate is a drug used for the treatment of gastritis and gastric ulcer, and has been proposed for use in the treatment of dry eye syndrome.
Especificaciones
Nombre del producto químico o material Gefarnate
CAS 51-77-4
Color Incolora
Notas de grado de pureza Research
Fórmula molecular C27H44O2
Cantidad 50 mg
Información de solubilidad DMSO : 31.25 mg/mL (78.00 mM; Need ultrasonic)
SMILES C/C(C)=C\CC/C(C)=C/CC/C(C)=C/CCC(OC/C=C(C)/CC/C=C(C)/C)=O
Peso molecular (g/mol) 400.64
Formula Weight (peso de la fórmula) 400.64
Porcentaje de pureza 98.0%
Grado Research
Forma física Liquid
Para utilizar con (aplicación) COVID-19-immunoregulation