missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GDF5 Polyclonal Antibody

Descripción
In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant mouse (rm) GDF-6 is observed, and less than 5% cross-reactivity with rmGDF-1, rmGDF-3, rmGDF-8, rmGDF-9, recombinant human GDF-11, and rhGDF-15 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
GDF-5 is expressed in long bones during embryonic development and postnatally in articular cartilage. Mutations in the GDF-5 gene have been implicated in Hunter-Thompson type dwarfism and in Grebe Syndrome, which is characterized by short stature, extra digits, and short and deformed extremities. The mature and functional form of GDF-5 is a homodimer of two 120 amino-acid polypeptide chain (monomers) linked by a single disulfide bond. Each GDF-5 monomer is expressed as the C-terminal part of a precursor polypeptide, which also contains a 27 amino acid signal peptide and a 354 amino acid propeptide. This precursor undergoes intracellular dimerization, and upon secretion it is processed by a furin-type protease.
Especificaciones
Especificaciones
| Antígeno | GDF5 |
| Aplicaciones | Immunohistochemistry (Frozen), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.2 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 5% trehalose and No Preservative |
| génica | GDF5 |
| N.º de referencia del gen | P43027 |
| Alias de gen | BDA1C; Bmp14; BMP-14; Bone morphogenetic protein 14; bp; brachypodism; brp; Cartilage-derived morphogenetic protein 1; cartilage-derived morphogenetic protein-1; CDMP1; Cdmp-1; GDF5; GDF-5; growth differentiation factor 5; growth/differentiation factor 5; H-GDF-5; LAP4; LAP-4; Lipopolysaccharide-associated protein 4; LPS-associated protein 4; OS5; radotermin; SYM1B; SYNS2 |
| Símbolos de los genes | GDF5 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?