Learn More
GB 23200.113-2018 Grupo A 117 Pesticidas 10 μg/ml en acetato de etilo, Dr. Ehrenstorfer
Especificaciones
Especificaciones
| Nombre del producto químico o material | (+)-trans-Permethrin∣2,4'-DDD∣2,4'-DDE∣2,4'-DDT∣4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Acephate∣Acetochlor∣Aclonifen∣Alachlor∣Allethrin∣alpha-Endosulfan∣alpha-HCH∣Anilofos∣Atrazine-desethyl∣beta-Endosulfan∣beta-HCH∣Bifenthrin∣Boscalid∣Bromacil∣Bromfenvinphos∣Bromophos-methyl∣Bromopropylate∣Bupirimate∣Carbophenothion∣Chlorthiophos∣Cycloate∣Cyflufenamid∣Cypermethrin∣delta-HCH∣Deltamethrin∣Dichlobenil∣Dichlofenthion∣Dichlorvos∣Dicloran∣Dicofol∣Dimethoate∣Dioxathion∣Ditalimfos∣Edifenphos∣Endrin∣EPN∣Epoxiconazole∣Ethalfluralin∣Ethoprophos∣Fenamidone∣Fenarimol∣Fenothiocarb∣Fensulfothion∣Fenthion∣Fenthion-sulfone∣Fenthion-sulfoxide∣Fenvalerate∣Flutolanil∣Fonofos∣Formothion∣Fosthiazate∣gamma-HCH∣Hexachlorobenzene∣Hexazinone∣Imazalil∣Iprobenfos∣Isofenphos∣Isofenphos-methyl∣Isoprocarb∣Isoxathion∣Kresoxim-methyl∣Mepanipyrim∣Mephosfolan∣Metalaxyl∣Methacrifos∣Methamidophos∣Metolachlor∣Metribuzin∣Mevinphos∣Molinate∣Monocrotophos∣Oxadiazon∣Oxyfluorfen∣Paraoxon-ethyl∣Paraoxon-methyl∣Parathion-ethyl∣Penconazole∣Phorate∣Phorate-sulfone∣Phorate-sulfoxide∣Piperonyl butoxide∣Piperophos∣Pirimicarb∣Pirimiphos-methyl∣Pretilachlor∣Profluralin∣Propazine∣Propetamphos∣Propoxur∣Pyrazophos∣Pyridaben∣Pyriproxyfen∣Quinalphos∣Quinoxyfen∣Sulfotep∣Tebuconazole∣Tebufenpyrad∣Tebupirimfos∣Terbufos∣Terbufos-sulfone∣Tetradifon∣Tetramethrin∣Thiobencarb∣Triadimefon∣Triadimenol∣Triallate∣Triazophos∣Tribufos∣Trifloxystrobin∣Vinclozolin |
| CAS | 51877-74-8, 53-19-0, 3424-82-6, 789-02-6, 72-54-8, 72-55-9 |
| Nombres de analitos o componentes | (+)-trans-Permethrin∣2,4'-DDD∣2,4'-DDE∣2,4'-DDT∣4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Acephate∣Acetochlor∣Aclonifen∣Alachlor∣Allethrin∣alpha-Endosulfan∣alpha-HCH∣Anilofos∣Atrazine-desethyl∣beta-Endosulfan∣beta-HCH∣Bifenthrin∣Boscalid∣Bromacil∣Bromfenvinphos∣Bromophos-methyl∣Bromopropylate∣Bupirimate∣Carbophenothion∣Chlorthiophos∣Cycloate∣Cyflufenamid∣Cypermethrin∣delta-HCH∣Deltamethrin∣Dichlobenil∣Dichlofenthion∣Dichlorvos∣Dicloran∣Dicofol∣Dimethoate∣Dioxathion∣Ditalimfos∣Edifenphos∣Endrin∣EPN∣Epoxiconazole∣Ethalfluralin∣Ethoprophos∣Fenamidone∣Fenarimol∣Fenothiocarb∣Fensulfothion∣Fenthion∣Fenthion-sulfone∣Fenthion-sulfoxide∣Fenvalerate∣Flutolanil∣Fonofos∣Formothion∣Fosthiazate∣gamma-HCH∣Hexachlorobenzene∣Hexazinone∣Imazalil∣Iprobenfos∣Isofenphos∣Isofenphos-methyl∣Isoprocarb∣Isoxathion∣Kresoxim-methyl∣Mepanipyrim∣Mephosfolan∣Metalaxyl∣Methacrifos∣Methamidophos∣Metolachlor∣Metribuzin∣Mevinphos∣Molinate∣Monocrotophos∣Oxadiazon∣Oxyfluorfen∣Paraoxon-ethyl∣Paraoxon-methyl∣Parathion-ethyl∣Penconazole∣Phorate∣Phorate-sulfone∣Phorate-sulfoxide∣Piperonyl butoxide∣Piperophos∣Pirimicarb∣Pirimiphos-methyl∣Pretilachlor∣Profluralin∣Propazine∣Propetamphos∣Propoxur∣Pyrazophos∣Pyridaben∣Pyriproxyfen∣Quinalphos∣Quinoxyfen∣Sulfotep∣Tebuconazole∣Tebufenpyrad∣Tebupirimfos∣Terbufos∣Terbufos-sulfone∣Tetradifon∣Tetramethrin∣Thiobencarb∣Triadimefon∣Triadimenol∣Triallate∣Triazophos∣Tribufos∣Trifloxystrobin∣Vinclozolin |
| Condiciones de envío | Room Temperature |
| Certificaciones/conformidad | ISO 17034 |
| Solvent or Matrix | Ethyl Acetate |
| Cantidad | 1.5 mL |
| Concentration or Composition (by Analyte or Components) | (+)-trans-Permethrin 10 μg/mL∣2,4'-DDD 10 μg/mL∣2,4'-DDE 10 μg/mL∣2,4'-DDT 10 μg/mL∣4,4'-DDD 10 μg/mL∣4,4'-DDE 10 μg/mL∣4,4'-DDT 10 μg/mL∣Acephate 10 μg/mL∣Acetochlor 10 μg/mL∣Aclonifen 10 μg/mL∣Alachlor 10 μg/mL∣Allethrin 10 μg/mL∣alpha-Endosulfan 10 μg/mL∣alpha-HCH 10 μg/mL∣Anilofos 10 μg/mL∣Atrazine-desethyl 10 μg/mL∣beta-Endosulfan 10 μg/mL∣beta-HCH 10 μg/mL∣Bifenthrin 10 μg/mL∣Boscalid 10 μg/mL∣Bromacil 10 μg/mL∣Bromfenvinphos 10 μg/mL∣Bromophos-methyl 10 μg/mL∣Bromopropylate 10 μg/mL∣Bupirimate 10 μg/mL∣Carbophenothion 10 μg/mL∣Chlorthiophos 10 μg/mL∣Cycloate 10 μg/mL∣Cyflufenamid 10 μg/mL∣Cypermethrin 10 μg/mL∣delta-HCH 10 μg/mL∣Deltamethrin 10 μg/mL∣Dichlobenil 10 μg/mL∣Dichlofenthion 10 μg/mL∣Dichlorvos 10 μg/mL∣Dicloran 10 μg/mL∣Dicofol 10 μg/mL∣Dimethoate 10 μg/mL∣Dioxathion 10 μg/mL∣Ditalimfos 10 μg/mL∣Edifenphos 10 μg/mL∣Endrin 10 μg/mL∣EPN 10 μg/mL∣Epoxiconazole 10 μg/mL∣Ethalfluralin 10 μg/mL∣Ethoprophos 10 μg/mL∣Fenamidone 10 μg/mL∣Fenarimol 10 μg/mL∣Fenothiocarb 10 μg/mL∣Fensulfothion 10 μg/mL∣Fenthion 10 μg/mL∣Fenthion-sulfone 10 μg/mL∣Fenthion-sulfoxide 10 μg/mL∣Fenvalerate 10 μg/mL∣Flutolanil 10 μg/mL∣Fonofos 10 μg/mL∣Formothion 10 μg/mL∣Fosthiazate 10 μg/mL∣gamma-HCH 10 μg/mL∣Hexachlorobenzene 10 μg/mL∣Hexazinone 10 μg/mL∣Imazalil 10 μg/mL∣Iprobenfos 10 μg/mL∣Isofenphos 10 μg/mL∣Isofenphos-methyl 10 μg/mL∣Isoprocarb 10 μg/mL∣Isoxathion 10 μg/mL∣Kresoxim-methyl 10 μg/mL∣Mepanipyrim 10 μg/mL∣Mephosfolan 10 μg/mL∣Metalaxyl 10 μg/mL∣Methacrifos 10 μg/mL∣Methamidophos 10 μg/mL∣Metolachlor 10 μg/mL∣Metribuzin 10 μg/mL∣Mevinphos 10 μg/mL∣Molinate 10 μg/mL∣Monocrotophos 10 μg/mL∣Oxadiazon 10 μg/mL∣Oxyfluorfen 10 μg/mL∣Paraoxon-ethyl 10 μg/mL∣Paraoxon-methyl 10 μg/mL∣Parathion-ethyl 10 μg/mL∣Penconazole 10 μg/mL∣Phorate 10 μg/mL∣Phorate-sulfone 10 μg/mL∣Phorate-sulfoxide 10 μg/mL∣Piperonyl butoxide 10 μg/mL∣Piperophos 10 μg/mL∣Pirimicarb 10 μg/mL∣Pirimiphos-methyl 10 μg/mL∣Pretilachlor 10 μg/mL∣Profluralin 10 μg/mL∣Propazine 10 μg/mL∣Propetamphos 10 μg/mL∣Propoxur 10 μg/mL∣Pyrazophos 10 μg/mL∣Pyridaben 10 μg/mL∣Pyriproxyfen 10 μg/mL∣Quinalphos 10 μg/mL∣Quinoxyfen 10 μg/mL∣Sulfotep 10 μg/mL∣Tebuconazole 10 μg/mL∣Tebufenpyrad 10 μg/mL∣Tebupirimfos 10 μg/mL∣Terbufos 10 μg/mL∣Terbufos-sulfone 10 μg/mL∣Tetradifon 10 μg/mL∣Tetramethrin 10 μg/mL∣Thiobencarb 10 μg/mL∣Triadimefon 10 μg/mL∣Triadimenol 10 μg/mL∣Triallate 10 μg/mL∣Triazophos 10 μg/mL∣Tribufos 10 μg/mL∣Trifloxystrobin 10 μg/mL∣Vinclozolin 10 μg/mL |