Learn More
Thermo Scientific™ Gas Chromatography SMART Syringe for TriPlus™ RSH SMART Autosampler
Tienda Thermo Scientific ProductosDescripción
Use the unique technology of Thermo Scientific™ gas chromatography SMART Syringes as a traceable usage-based approach to gas chromatography (gas chromatography) syringe management, resulting in increased reliability, instrument up-time, confidence in the results, and full traceability.
SMART Technology Benefits:
- Gain higher confidence in analytical results
- Ensure error-free usage of syringes through automatic identification
- Support GLP accreditation with usage history traceability
- Optimize consumption by monitoring usage conditions and lifetime
- Facilitate syringe management with a color-coded health notification dashboard integrated in Chromeleon™ CDS
Especificaciones
Especificaciones
| Material | Borosilicate Glass (Barrel) |
| Tipo de producto | Syringe |
| Volumen (métrico) | 10 μL |
| Graduated | Yes |