Learn More
Tocris Bioscience™ Ganaxolone
Descripción
Potent positive allosteric modulator of GABAA receptors. Enhances GABA-evoked chloride currents in Xenopus oocytes expressing GABAA receptors (EC50 values are 94, 122 and 213 nM for α2β1γ2L, α3β1γ2L and α1β1γ2L receptors respectively). Exerts anticonvulsive effects in a broad range of animal seizure models.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Ganaxolone |
| CAS | 38398-32-2 |
| Cantidad | 50 mg |
| Fórmula molecular | C22H36O2 |
| Formula Weight (peso de la fórmula) | 332.53 |
| Pureza | >98% |
| Peso molecular | 332.53 |