Learn More
Tocris Bioscience™ Galidesivir dihydrochloride
Descripción
Galidesivir is a viral RNA-dependent RNA polymerase (RdRP) inhibitor and broad spectrum antiviral nucleotide. In vitro, galidesivir displays antiviral activity against a range of RNA viruses, including bunyaviruses, arenaviruses, paramyxoviruses, coronaviruses, and flaviviruses (EC50 values range from 3 μM to >100 μM). Galidesivir protects against Ebola and Marburg viral infection in animal models.
Especificaciones
Especificaciones
| Nombre del producto químico o material | (2 S,3 S,4 R,5 R)-2-(4-Amino-5 H-pyrrolo[3,2-d]pyrimidin-7-yl)-5-(hydroxymethyl)-3,4-pyrrolidinediol dihydrochloride |
| CAS | 1373208-51-5 |
| Cantidad | 2 mg |
| Objetivo | RNA/DNA Polymerase Inhibitors |
| Fórmula molecular | C11H15N5O30.2 HCl |
| Pureza | 0.98 |