missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Galectin 8 Polyclonal Antibody
Descripción
Immunogen sequence: PFVGTIPDQL DPGTLIVIRG HVPSDADRFQ VDLQNGSSMK PRADVAFHFN PRFKRAGCIV CNTLINEKWG REEITYDTPF KREKSFEIVI MVLKDKFQVA VNGKHTLLYG HRIGPEKIDT LGIYGKVNIH SIGFSFSSDL QSTQASS Highest antigen sequence indentity to the following orthologs: Mouse - 78%, Rat - 80%.
This gene encodes a member of the galectin family. Galectins are beta-galactoside-binding animal lectins with conserved carbohydrate recognition domains. The galectins have been implicated in many essential functions including development, differentiation, cell-cell adhesion, cell-matrix interaction, growth regulation, apoptosis, and RNA splicing. This gene is widely expressed in tumoral tissues and seems to be involved in integrin-like cell interactions. Alternatively spliced transcript variants encoding different isoforms have been identified.
Especificaciones
Especificaciones
| Antígeno | Galectin 8 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | Lgals8 |
| N.º de referencia del gen | O00214 |
| Alias de gen | 1200015E08Rik; 30 kDa S-type lectin; AI326142; D13Ertd524e; Gal-8; galectin 8; galectin-8; galectin-8g; galectin-8L; lectin, galactose binding, soluble 5; lectin, galactose binding, soluble 8; lectin, galactoside binding soluble 8; lectin, galactoside-binding, soluble, 8; LGALS8; Lgals-8; PCTA1; PCTA-1; Po66 carbohydrate binding protein; po66 carbohydrate-binding protein; Po66-CBP; prostate carcinoma tumor antigen 1; RL-30; RP11-385F5.1 |
| Símbolos de los genes | Lgals8 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?