Learn More
GAL-021, MedChemExpress
Código de producto. 16459886
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16459886 50 mg 50 miligramos
16453358 10 mg 10 miligramos
16416087 5 mg 5 miligramos
16414606 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16459886 Proveedor Medchem Express N.º de proveedor HY101422
MedChemExpress GAL-021 is a potent BKCa-channel blocker. GAL-021 inhibits KCa1.1 in GH3 cells. GAL-021 is a novel breathing control modulator that is based on selective modification of the almitrine pharmacophore. GAL-021 increases minute ventilation in rats and non-human primates.
GAL-021 is a potent BKCa-channel blocker. GAL-021 inhibits KCa1.1 in GH3 cells. GAL-021 is a novel breathing control modulator that is based on selective modification of the almitrine pharmacophore. GAL-021 increases minute ventilation in rats and non-human primates.
Especificaciones
Nombre del producto químico o material GAL-021
Forma física Solid
CAS 1380341-99-0
Color Blanco
Cantidad 50 mg
Notas de grado de pureza Research
Fórmula molecular C11H22N6O
Para utilizar con (aplicación) COVID-19-immunoregulation
Información de solubilidad DMSO : ≥ 30 mg/mL (117.96 mM)
SMILES CCCNC1=NC(N(OC)C)=NC(NCCC)=N1
Peso molecular (g/mol) 254.33
Formula Weight (peso de la fórmula) 254.33
Porcentaje de pureza 98.0%