Learn More
Tocris Bioscience™ G-36
Descripción
G-36 is an antagonist of GPER; selectively inhibits estrogen-mediated activation of PI3-K by GPER, but not by ERα. Also inhibits estrogen-mediated calcium mobilization (IC50 = 112 nM). Structural analog of G-1 (Cat. No. 3577).
Especificaciones
Especificaciones
| Nombre del producto químico o material | (±)-(3aR*,4S*,9bS*)-4-(6-Bromo-1,3-benzodioxol-5-yl)-3a,4,5,9b-tetrahydro-8-(1-methylethyl)-3H-cyclopenta[c]quinoline |
| CAS | 1392487-51-2 |
| Cantidad | 10 mg |
| Objetivo | Estrogen and Related Receptor Antagonists |
| Fórmula molecular | C22H22BrNO2 |
| Pureza | 99% |