Learn More
Tocris Bioscience™ G 28UCM
FASN inhibitor
Marca: Tocris Bioscience™ 5446/10
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
G 28UCM is a FASN inhibitor. Inhibits breast cancer cell proliferation in vitro (IC50 values are 21 - 79 μM), including cell lines resistant to anti-HER2 therapies. Blocks HER2 signaling and induces apoptosis. Inhibits growth of breast cancer xenografts in mice without causing anorexia.Especificaciones
| 3-(3,4,5-Trihydroxybenzoyloxy)naphthalen-1-yl 3,4,5-trihydroxybenzoate | |
| 1094451-90-7 | |
| 10 mg |