Learn More
Tocris Bioscience™ FX 1
Descripción
FX 1 is a Bcl-6 inhibitor (Kd = 3 μM); exhibits 10-fold greater potency than endogenous Bcl-6 corepressors. Disrupts formation of Bcl-6 repression complex. Induces regression of established tumors in mice bearing DLBCL xenograft.
Especificaciones
Especificaciones
| Nombre del producto químico o material | (5Z)-5-(5-Chloro-1,2-dihydro-2-oxo-3H-indol-3-ylidene)-4-oxo-2-thioxo-3-thiazolidinepropanoic acid |
| CAS | 1426138-42-2 |
| Cantidad | 5 mg |
| Fórmula molecular | C14H9ClN2O4S2 |
| Formula Weight (peso de la fórmula) | 368.82 |
| Pureza | 98% |