Learn More
Futibatinib, MedChemExpress
Código de producto. 16485907
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
5 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16485907 5 mg 5 miligramos
16479726 50 mg 50 miligramos
16463178 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16485907 Proveedor Medchem Express N.º de proveedor HY1008185mg
MedChemExpress Futibatinib (TAS-120) is an orally bioavailable, highly selective, and irreversible FGFR inhibitor, with IC50s of 3.9, 1.3, 1.6, and 8.3 nM for FGFR 1-4, respectively. Futibatinib inhibits mutant and wild-type FGFR2 with similar IC50s (wild-type FGFR2=0.9 nM; V5651=1-3 nM; N550H=3.6 nM; E566G=2.4 nM).
Futibatinib (TAS-120) is an orally bioavailable, highly selective, and irreversible FGFR inhibitor, with IC50s of 3.9, 1.3, 1.6, and 8.3 nM for FGFR 1-4, respectively. Futibatinib inhibits mutant and wild-type FGFR2 with similar IC50s (wild-type FGFR2=0.9 nM
Especificaciones
Nombre del producto químico o material Futibatinib
CAS 1448169-71-8
Color Off-White
Notas de grado de pureza Research
Fórmula molecular C22H22N6O3
Cantidad 5 mg
Sinónimo TAS-120
Información de solubilidad DMSO : ≥ 29 mg/mL (69.30 mM)
SMILES O=C(C=C)N(C1)CC[C@@H]1N(N=C2C#CC3=CC(OC)=CC(OC)=C3)C4=C2C(N)=NC=N4
Peso molecular (g/mol) 418.45
Formula Weight (peso de la fórmula) 418.45
Porcentaje de pureza 98.02%
Grado Research
Forma física Powder
Para utilizar con (aplicación) Cancer-Kinase/protease