Learn More
Tocris Bioscience™ Furamidine dihydrochloride
Descripción
Furamidine dihydrochloride is a selective protein arginine methyltransferase 1 (PRMT1) inhibitor (IC50 = 9.4 μM). Exhibits selectivity over PRMT5, PRMT6 and CARM1 (IC50 values are 166, 283 and >400 μM respectively). Inhibits cell proliferation in leukemia cell lines. Cell-permeable.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 4,4'-(2,5-Furandiyl)bis-benzenecarboximidamide dihydrochloride |
| CAS | 55368-40-6 |
| Intervalo de porcentaje de ensayo | >98% |
| Cantidad | 5 mg |
| Fórmula molecular | C18H16N4O.2HCl |
| Formula Weight (peso de la fórmula) | Observed MW: 377.27 |