Learn More
Tocris Bioscience™ FTY 720
Potent S1P receptor agonist; also immunosuppressant
Marca: Tocris Bioscience™ 6176/50
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
FTY 720 is a S1P receptor agonist; undergoes phosphorylation by sphingosine kinase 2, which then binds S1P1,3,4 and 5 receptors (EC50 values are 0.3 - 0.6 nM for S1P1, 4 and 5 and ∽3 nM for S1P3). Immunosuppresant; sequesters lymphocytes in lymph nodes, reducing autoimmune response. Suppresses the development of EAE (an in vivo model of MS). Orally bioavailable and blood-brain barrier permeable.Especificaciones
| Potent S1P Receptor Agonist, also Immunosuppressant | |
| 162359-56-0 | |
| 343.93g/mol | |
| Store at -20°C | |
| 50 mg |
| Sphingosine-1-phosphate Receptor | |
| C19H33NO2.HCl | |
| Fingolimod, Gilenya | |
| Sphingosine-1-phosphate Receptor Agonists |
Título del producto
For Research Use Only