missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FTO Monoclonal antibody specifically detects FTO in Human, Mouse, Pig samples. It is validated for IHC, WB, ELISA
Specifications
Specifications
| Antígeno | FTO |
| Aplicaciones | Immunohistochemistry, Western Blot, ELISA |
| Clasificación | Monoclonal |
| Clon | 1E8B1 |
| Concentración | 1000 μg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.02% sodium azide and 50% glycerol pH 7.3. |
| N.º de referencia del gen | BC030266 |
| Especie del huésped | Mouse |
| Método de purificación | Protein A purification |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?