Learn More
Fraxin, MedChemExpress
Código de producto. 16429028
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
Tamaño de la unidad:
10 miligramos
25 miligramos
5 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16429028 10 mg 10 miligramos
16413886 25 mg 25 miligramos
16401448 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16429028 Supplier Medchem Express Supplier No. HYN0579
This item is not returnable. View return policy
MedChemExpress Fraxin isolated from Acer tegmentosum, F. ornus or A. hippocastanum, is a glucoside of fraxetin and reported to exert potent anti-oxidative stress action, anti-inflammatory and antimetastatic properties. Fraxin shows its antioxidative effect through inhibition of cyclo AMP phosphodiesterase enzyme.
Fraxin isolated from Acer tegmentosum, F. ornus or A. hippocastanum, is a glucoside of fraxetin and reported to exert potent anti-oxidative stress action, anti-inflammatory and antimetastatic properties. Fraxin shows its antioxidative effect through inhibition of cyclo AMP phosphodiesterase enzyme.
Specifications
Nombre del producto químico o material Fraxin
CAS 524-30-1
Color Light Yellow
Notas de grado de pureza Research
Fórmula molecular C16H18O10
Cantidad 10 mg
Sinónimo Fraxoside
Información de solubilidad DMSO : 250 mg/mL (675.11 mM; Need ultrasonic)
SMILES OC1=C(OC)C=C(C=C2)C(OC2=O)=C1O[C@@H]([C@@H]([C@@H](O)[C@@H]3O)O)O[C@@H]3CO
Peso molecular (g/mol) 370.31
Formula Weight (peso de la fórmula) 370.31
Porcentaje de pureza 98.55%
Grado Research
Forma física Solid
Para utilizar con (aplicación) COVID-19-immunoregulation