Learn More
Tocris Bioscience™ FPS ZM1
Descripción
FPS ZM1 is a high affinity antagonist of the receptor for advanced glycation end products (RAGE) (Ki = 25 nM). Blocks Aβ binding to the V domain of RAGE and inhibits Aβ40- and Aβ42-induced cellular stress in RAGE-expressing cells in vitro and in the mouse brain in vivo. Blocks β-secretase activity, Aβ production, microglia activation and neuroinflammation in vivo. BBB permeable.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 4-Chloro-N-cyclohexyl-N-(phenylmethyl)benzamide |
| CAS | 945714-67-0 |
| Intervalo de porcentaje de ensayo | >99% |
| Cantidad | 10 mg |
| Fórmula molecular | C20H22ClNO |
| Formula Weight (peso de la fórmula) | Observed MW: 327.85 |