missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXO4 Polyclonal Antibody

Descripción
Recommended positive controls: Cos-7. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FOXO4 (Forkhead box O4) is encoded by the FOXO4 gene located on the X chromosome. It is also known as AFX1 or MLLT7 (Myeloid/Lymphoid or Mixed-Lineage Leukemia (Trithorax Homolog, Translocated To, 7) and is a member of the O class of forkhead/ winged helix family of transcription factors along with FOXO1, FOXO3 and FOXO6. FOXO4 is abundantly expressed in skeletal muscle and adipose tissue. FOXO4 binds to the Insulin Responsive Element (IRE) and activates downstream targets of Insulin/IGF1 signaling such as IGFBP1. However, the activation of another downstream target of the Insulin/IGF1 pathway, Akt, leads to phosphorylation of FOXO4 on specific Ser/Thr residues. This phosphorylation mediates binding of FOXO4 to 14-3-3 and eventual exit of FOXO4 from the nucleus.
Especificaciones
Especificaciones
| Antígeno | FOXO4 |
| Aplicaciones | Immunoprecipitation, Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA and 15mM sodium azide; pH 7.4 |
| génica | FOXO4 |
| N.º de referencia del gen | P98177 |
| Alias de gen | AFX; Afx1; Afxh; AFXzeta; Fkhr3; Fork head domain transcription factor AFX1; forkhead box O4; forkhead box protein O4; forkhead protein; FOXO4; FOXO4b; MGC120490; MLLT7; myeloid/lymphoid or mixed lineage-leukemia translocation to 7 homolog; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 7; OTTHUMP00000023497; RGD1561201 |
| Símbolos de los genes | FOXO4 |
| Especie del huésped | Rabbit |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?