missing translation for 'onlineSavingsMsg'
Learn More

FOXJ3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16860855 Shop All Proteintech Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Cantidad:
20 μL
150 μL
missing translation for 'unitSize'
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
16860855 150 μL 150 microlitros
16870855 20 μL 20 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 16860855 missing translation for 'mfr' Proteintech missing translation for 'supplierNo' 212401AP150UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Transcriptional activator of MEF2C involved in the regulation pf adult muscle fiber type identity and skeletal muscle regeneration.

Specifications

Antígeno FOXJ3
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.35 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica FOXJ3
N.º de referencia del gen Q8BUR3, Q9UPW0
Alias de gen forkhead box J3, Forkhead box protein J3, FOXJ3, KIAA1041
Símbolos de los genes Foxj3
Especie del huésped Rabbit
Inmunógeno FOXJ3 Fusion Protein Ag15732
Método de purificación Antigen Affinity Chromatography
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 22887, 230700, 313554
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.