Learn More
Abeba Food Trax Safety Shoes
Descripción
- ESD-Compliant Safety Shoes Designed for Use in ESD-Sensitive Environments
- S3 Classification Provides Workplace Safety Protection
- Compliant with DIN EN 61340 ESD Requirements
- Composite Toe Cap Helps Protect Against Impact and Compression Hazards
- Antistatic Design Helps Reduce Electrostatic Charge Buildup
- Black Upper Provides a Professional Appearance
- Available in Sizes 36 to 50
Especificaciones
Especificaciones
| Color | Black |
| Material | Microfiber |
| Tamaño | 42 |
| Material del dedo del pie | Composite |
| Desechables | Reusable |
| Sexo | Unisex |
| Ajuste | Standard Width |
| Sin látex | Yes |
| Waterproof | Water Resistant |
| EN ISO 20345 | S3 |
| Mostrar más |