Learn More
Tocris Bioscience™ Flutax 1
Descripción
Flutax 1 is a a fluorescent taxol derivative that binds to the taxol microtubule binding site with high affinity (Ka ∼ 107M-1). Can be used for direct imaging of the microtubule cytoskeleton. Absorption, fluorescence and fluorescence decay of Flutax 1 in solution are pH sensitive. Excitation maximum ∼ 495 nm; emission maximum ∼ 520 nm.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Flutax 1 |
| CAS | 191930-58-2 |
| Cantidad | 1 mg |
| Fórmula molecular | C71H66N2O21 |
| Formula Weight (peso de la fórmula) | 1283.3 |
| Pureza | >98% |
| Peso molecular | 1283.302 |