Learn More
Tocris Bioscience™ Fluorescein Tyramide
Descripción
Fluorescein Tyramide is a green fluorescent reagent widely used for tyramide signal amplification (TSA) in IHC, ICC, FISH and multicolor FISH. HRP catalyzes localized deposition of multiple tyramide molecules (catalyzed reporter deposition, CARD), binding the fluorescein tyramide to adjacent tyrosines to enhance fluorescent signal.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 3 ',6 '-Dihydroxy-N-(4-hydroxyphenylethyl)-3-oxo-3 H-spiro[isobenzofuran-1,9 '-xanthene]-5(6)-carboxamide |
| CAS | 210236-90-1 |
| Cantidad | 1 mg |
| Objetivo | Tyramide Signaling Amplification/TSA |
| Fórmula molecular | C29H21NO7 |
| Formula Weight (peso de la fórmula) | 2020 |
| Pureza | 95% |